ENA333557624 Enamine

(3-methoxy-4-methylphenyl)methanol

  • MDL number MFCD06797828
  • Cas Number : 4685-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70691
Availability details for
Quantity
  • Availability
  • Add to Cart