ENA333557631 Enamine

2,2,2-trifluoroethanethioamide

  • MDL number MFCD02093844
  • Cas Number : 421-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81703

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-81703
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart