ENA333557643 Enamine

2-[2-(hydroxymethyl)pyrrolidin-1-yl]acetic acid

  • MDL number MFCD14695322
  • Cas Number : 155681-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81721
Availability details for
Quantity
  • Availability
  • Add to Cart