ENA333557653 Enamine

1-(naphthalen-2-ylmethyl)piperazine

  • MDL number MFCD00435480
  • Cas Number : 61187-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81737

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81737
Availability details for
Quantity
  • Availability
  • Add to Cart