ENA333557656 Enamine

1-(3-methylthiophen-2-yl)-2-phenylethan-1-amine

  • MDL number MFCD00908940
  • Cas Number : 143380-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81741
Availability details for
Quantity
  • Availability
  • Add to Cart