ENA333557661 Enamine

N-ethyl-4-iodobenzamide

  • MDL number MFCD03703872
  • Cas Number : 113948-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81748
Availability details for
Quantity
  • Availability
  • Add to Cart