ENA333557663 Enamine

4-bromo-N-(pyridin-2-yl)benzamide

  • MDL number MFCD00447937
  • Cas Number : 14547-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81750

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81750
Availability details for
Quantity
  • Availability
  • Add to Cart