ENA333557682 Enamine

2-(3-Chloro-5-trifluoromethyl-pyridin-2-ylamino)-ethanol

  • MDL number MFCD10568236
  • Cas Number : 1053656-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03187

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03187
Availability details for
Quantity
  • Availability
  • Add to Cart