ENA333557683 Enamine

N1-[4-(trifluoromethoxy)phenyl]benzene-1,4-diamine

  • MDL number MFCD10690539
  • Cas Number : 1152561-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-81780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81780
Availability details for
Quantity
  • Availability
  • Add to Cart