ENA333557690 Enamine

N-benzyl-2-(methylamino)acetamide

  • MDL number MFCD02266689
  • Cas Number : 74963-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81792
Availability details for
Quantity
  • Availability
  • Add to Cart