ENA333557691 Enamine

methyl 2-oxo-2-(2-oxocyclopentyl)acetate

  • MDL number MFCD06644285
  • Cas Number : 63053-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27846

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27846
Availability details for
Quantity
  • Availability
  • Add to Cart