ENA333557699 Enamine

pyrazin-2-ylmethanol

  • MDL number MFCD01693938
  • Cas Number : 6705-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81801
Availability details for
Quantity
  • Availability
  • Add to Cart