ENA333557701 Enamine

1-phenyl-2-(trifluoromethoxy)ethan-1-one

  • MDL number MFCD17976633
  • Cas Number : 104159-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81806
Availability details for
Quantity
  • Availability
  • Add to Cart