ENA333557708 Enamine

2-fluoro-5-(2-oxoimidazolidin-1-yl)benzoic acid

  • MDL number MFCD12158659
  • Cas Number : 1183572-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 319...321
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81816

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81816
Availability details for
Quantity
  • Availability
  • Add to Cart