ENA333557709 Enamine

5-{[2-(trifluoromethyl)phenyl]methylidene}-1,3-thiazolidine-2,4-dione

  • Cas Number : 292172-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81817
Availability details for
Quantity
  • Availability
  • Add to Cart