ENA333557711 Enamine

2-[3-(1H-1,2,4-triazol-1-yl)propoxy]benzonitrile

  • MDL number MFCD05986830
  • Cas Number : 695158-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81819
Availability details for
Quantity
  • Availability
  • Add to Cart