ENA333557714 Enamine

3-[2-(propan-2-yl)-1H-imidazol-1-yl]piperidine

  • MDL number MFCD16076820
  • Cas Number : 1249887-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71193
Availability details for
Quantity
  • Availability
  • Add to Cart