ENA333557721 Enamine

methyl({1-[4-methyl-2-(4-methylphenyl)-1,3-thiazol-5-yl]ethyl})amine

  • MDL number MFCD13332425
  • Cas Number : 1291467-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72130
Availability details for
Quantity
  • Availability
  • Add to Cart