ENA333557733 Enamine

N-ethyl-4-methoxyaniline

  • MDL number MFCD03210676
  • Cas Number : 104-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33075
Availability details for
Quantity
  • Availability
  • Add to Cart