ENA333557736 Enamine

2-[1-(propan-2-yl)-1H-pyrazol-3-yl]acetonitrile

  • MDL number MFCD18262432
  • Cas Number : 1260659-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74580
Availability details for
Quantity
  • Availability
  • Add to Cart