ENA333557737 Enamine

2-chloroquinoline-4-carbonitrile

  • MDL number MFCD02041240
  • Cas Number : 4366-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153...155
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74592

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74592
Availability details for
Quantity
  • Availability
  • Add to Cart