ENA333557743 Enamine

5,6-dihydro-2H-thiopyran-3-carboxylic acid

  • MDL number MFCD19227910
  • Cas Number : 172214-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75216
Availability details for
Quantity
  • Availability
  • Add to Cart