ENA333557745 Enamine

tert-butyl N-(3-methylpiperidin-4-yl)carbamate

  • MDL number MFCD09701320
  • Cas Number : 866538-96-7
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75493
Availability details for
Quantity
  • Availability
  • Add to Cart