ENA333557746 Enamine

methyl 2-amino-2-cyclopropylpropanoate hydrochloride

  • MDL number MFCD19686207
  • Cas Number : 1333675-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35561

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35561
Availability details for
Quantity
  • Availability
  • Add to Cart