ENA333557747 Enamine

4-(3-bromophenyl)-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD12138992
  • Cas Number : 1309887-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75549
Availability details for
Quantity
  • Availability
  • Add to Cart