ENA333557749 Enamine

6-methylimidazo[2,1-b][1,3]thiazole

  • MDL number MFCD04610181
  • Cas Number : 3835-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-17423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17423
Availability details for
Quantity
  • Availability
  • Add to Cart