ENA333557750 Enamine

3-(1H-imidazol-1-yl)phenol

  • MDL number MFCD19686220
  • Cas Number : 10041-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75587

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75587
Availability details for
Quantity
  • Availability
  • Add to Cart