ENA333557751 Enamine

6-bromo-1-ethyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD11148032
  • Cas Number : 946112-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81861

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81861
Availability details for
Quantity
  • Availability
  • Add to Cart