ENA333557753 Enamine

2-(1H-indazol-1-yl)ethan-1-ol

  • MDL number MFCD16739318
  • Cas Number : 1236127-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81863
Availability details for
Quantity
  • Availability
  • Add to Cart