ENA333557756 Enamine

2-methyl-4-(2-methylphenyl)pyrrolidine

  • MDL number MFCD19686392
  • Cas Number : 1333969-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81866
Availability details for
Quantity
  • Availability
  • Add to Cart