ENA333557760 Enamine

2-amino-1-(3,4-dimethoxyphenyl)ethan-1-ol hydrochloride

  • MDL number MFCD00798887
  • Cas Number : 15471-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81873

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81873
Availability details for
Quantity
  • Availability
  • Add to Cart