ENA333557762 Enamine

4-(aminomethyl)thian-4-ol

  • MDL number MFCD11040538
  • Cas Number : 879514-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65...67
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81882

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81882
Availability details for
Quantity
  • Availability
  • Add to Cart