ENA333557763 Enamine

4-fluoro-2-(piperidin-4-yl)-1H-1,3-benzodiazole

  • MDL number MFCD12150240
  • Cas Number : 1184760-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75630
Availability details for
Quantity
  • Availability
  • Add to Cart