ENA333557764 Enamine

bis(thiophen-2-yl)methanamine hydrochloride

  • MDL number MFCD19686395
  • Cas Number : 1333517-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81886
Availability details for
Quantity
  • Availability
  • Add to Cart