ENA333557770 Enamine

5-(4-chlorobenzoyl)-1,3-dioxaindane

  • MDL number MFCD11545106
  • Cas Number : 1096987-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81892
Availability details for
Quantity
  • Availability
  • Add to Cart