ENA333557775 Enamine

3-(4-methylphenyl)pentanoic acid

  • MDL number MFCD08282293
  • Cas Number : 57960-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81900
Availability details for
Quantity
  • Availability
  • Add to Cart