ENA333557777 Enamine

2-[(cyclopropylmethyl)(methyl)amino]-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD17114543
  • Cas Number : 1334146-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81903
Availability details for
Quantity
  • Availability
  • Add to Cart