ENA333557778 Enamine

5-(1-chloroethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole

  • MDL number MFCD10693627
  • Cas Number : 1152532-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39378
Availability details for
Quantity
  • Availability
  • Add to Cart