ENA333557792 Enamine

ethyl 5-methoxy-3-oxopentanoate

  • MDL number MFCD03844433
  • Cas Number : 104629-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81925
Availability details for
Quantity
  • Availability
  • Add to Cart