ENA333557793 Enamine

ethyl oxolane-3-carboxylate

  • MDL number MFCD06204767
  • Cas Number : 139172-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81926
Availability details for
Quantity
  • Availability
  • Add to Cart