ENA333557794 Enamine

ethyl 3-(benzyloxy)propanoate

  • MDL number MFCD11554979
  • Cas Number : 127113-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81927

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81927
Availability details for
Quantity
  • Availability
  • Add to Cart