ENA333557797 Enamine

N-[(2-aminophenyl)methyl]-N-methylthiophene-2-carboxamide

  • MDL number MFCD12413408
  • Cas Number : 1155611-16-7
  • Molecular Weight : 246.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-81930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81930
Availability details for
Quantity
  • Availability
  • Add to Cart