ENA333557801 Enamine

1-(2-methoxyethoxy)-3,3-dimethylbutan-2-one

  • MDL number MFCD12062255
  • Cas Number : 1158069-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81936

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81936
Availability details for
Quantity
  • Availability
  • Add to Cart