ENA333557802 Enamine

1-(2,4-difluorophenyl)propan-1-ol

  • MDL number MFCD12787299
  • Cas Number : 1247998-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81937
Availability details for
Quantity
  • Availability
  • Add to Cart