ENA333557805 Enamine

methyl 2-[(cyanomethyl)amino]acetate

  • MDL number MFCD11150527
  • Cas Number : 1021072-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81941

Safety Information

signalword Danger
pcodes P264,P270,P301+P310,P330,P405,P501
hcodes T
MSDS https://www.enaminestore.com/catalog/EN300-81941
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart