ENA333557823 Enamine

6-(pyrrolidine-1-carbonyl)pyridine-2-carboxylic acid

  • MDL number MFCD12137832
  • Cas Number : 1153759-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81967
Availability details for
Quantity
  • Availability
  • Add to Cart