ENA333557825 Enamine

3-(2-furyl)-1-(4-methylphenyl)-1H-pyrazol-5-amine

  • MDL number MFCD08442297
  • Cas Number : 1333887-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81969

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81969
Availability details for
Quantity
  • Availability
  • Add to Cart