ENA333557836 Enamine

methyl 1,2,3,4-tetrahydroquinoline-5-carboxylate hydrochloride

  • MDL number MFCD09026774
  • Cas Number : 1334147-75-9
  • Molecular Weight : 227.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-81985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81985
Availability details for
Quantity
  • Availability
  • Add to Cart