ENA333557838 Enamine

3-amino-2,5-difluorobenzoic acid

  • MDL number MFCD17276578
  • Cas Number : 1333537-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81987

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81987
Availability details for
Quantity
  • Availability
  • Add to Cart