ENA333557842 Enamine

2-(1,2-oxazol-3-yl)acetonitrile

  • MDL number MFCD16629913
  • Cas Number : 55242-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77189
Availability details for
Quantity
  • Availability
  • Add to Cart